Learn More
ZZEF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556856
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DYFFLEVQKR FDGDELTTDE RIRSLAQRWQ PSKSLRLEEQ SAKAVDTDMI ILPCLSRPAR CDQATAESNP VTQKLI Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
ZZEF1 gene ontology annotations related to this gene include calcium ion binding; zinc ion binding.Specifications
ZZEF1 | |
Polyclonal | |
Unconjugated | |
ZZEF1 | |
FLJ10821; KIAA0399; zinc finger ZZ-type and EF-hand domain containing 1; zinc finger ZZ-type and EF-hand domain-containing protein 1; zinc finger, ZZ-type with EF hand domain 1; zinc finger, ZZ-type with EF-hand domain 1; ZZEF1; ZZZ4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23140 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43149 | |
ZZEF1 | |
Recombinant protein corresponding to Human ZZEF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only