accessibility menu, dialog, popup

UserName

CAS RN 106-49-0

IUPAC Name: 4-methylaniline
Synonyms: toluidine naphtol AS-kgll benzenamine, ar-methyl CI azoic coupling component 107 ccris 598 p-toluidyna C.I. azoic coupling component 107 rcra waste number U353 naphtol AS-KG
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: RZXMPPFPUUCRFN-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (4)
  • (4)
  • (1)