missing translation for 'onlineSavingsMsg'
Learn More

p-Toluidine, 99%, crystalline molten mass

Product Code. 12694967 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Glass Bottle
HDPE Bottle
Metal drum
Unit Size:
1kg
250g
25kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12694967 5 kg HDPE Bottle 5kg
10024742 250 g Glass Bottle 250g
10128022 1 kg Glass Bottle 1kg
12624977 25 kg Metal drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 12694967 Supplier Thermo Scientific Acros Supplier No. 376350050
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 106-49-0 | C7H9N | 107.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-49-0
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
InChI Key RZXMPPFPUUCRFN-UHFFFAOYSA-N
Synonym p-toluidine, 4-toluidine, p-tolylamine, 4-aminotoluene, benzenamine, 4-methyl, 4-methylbenzenamine, p-methylaniline, p-methylbenzenamine, p-toluidin, p-aminotoluene
PubChem CID 7813
ChEBI CHEBI:37825
IUPAC Name 4-methylaniline
SMILES CC1=CC=C(C=C1)N

Specifications

Melting Point 41.0°C to 46.0°C
Color White to Brown
Boiling Point 200.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging HDPE Bottle
Linear Formula CH3C6H4NH2
Quantity 5 kg
Merck Index 15,9696
Solubility Information Solubility in water: 1.1 g/100mL. Other solubilities: freely soluble in diluted acids, freely soluble in alcohol, ether, acetone, carbon, disulfide, methanol and oils, 156% in ethanol
Formula Weight 107.16
Percent Purity 99%
Physical Form Crystalline Mass or Crystals
Assay 99%
Chemical Name or Material p-Toluidine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.