accessibility menu, dialog, popup

UserName

CAS RN 1075-89-4

IUPAC Name: 8-azaspiro[4.5]decane-7,9-dione
Synonyms: azaspirodecanedione
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: YRTHJMQKDCXPAY-UHFFFAOYSA-N
SMILES: O=C1CC2(CCCC2)CC(=O)N1
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)