accessibility menu, dialog, popup

UserName

H3COPSOCH3OSCH3O

CAS RN 115-90-2

IUPAC Name: O,O-diethyl O-4-methanesulfinylphenyl phosphorothioate
Synonyms: fensulfothion VUAgT 108 caswell no. 343 daconit chemagro 25141 VUAgT 96 epa pesticide chemical code 032701 terracur P dasanit
Molecular Weight (g/mol): 308.35
Molecular Formula: C11H17O4PS2
InChi Key: XDNBJTQLKCIJBV-UHFFFAOYNA-N
SMILES: CCOP(=S)(OCC)OC1=CC=C(C=C1)S(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results