accessibility menu, dialog, popup

UserName

CH3OOHNOCH3HCl

CAS RN 120013-39-0

IUPAC Name: 5,6-dimethoxy-2-[(piperidin-4-yl)methyl]-2,3-dihydro-1H-inden-1-one hydrochloride
Synonyms: 5,6-dimethoxy-2-(piperidin-4-ylmethyl)-2,3-dihydroinden-1-one hydrochloride
Molecular Weight (g/mol): 325.83
Molecular Formula: C17H24ClNO3
InChi Key: WOZXDQQCMZIQEG-UHFFFAOYNA-N
SMILES: Cl.COC1=C(OC)C=C2C(=O)C(CC3CCNCC3)CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results