accessibility menu, dialog, popup

UserName

CAS RN 120786-18-7

CH3H3CNH2ONH(E/Z)

CAS RN 120786-18-7

IUPAC Name: 1-amino-13-ethylidene-11-methyl-6-azatricyclo[7.3.1.0²,⁷]trideca-2(7),3,10-trien-5-one
Synonyms: 1-amino-13-ethylidene-11-methyl-6-azatricyclo[7.3.1.0²,⁷]trideca-2(7),3,10-trien-5-one
Molecular Weight (g/mol): 242.32
Molecular Formula: C15H18N2O
InChi Key: ZRJBHWIHUMBLCN-UHFFFAOYNA-N
SMILES: CC=C1C2CC3=C(C=CC(=O)N3)C1(N)CC(C)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results