CAS RN 122320-73-4
CAS RN 122320-73-4
IUPAC Name:
5-[(4-{2-[methyl(pyridin-2-yl)amino]ethoxy}phenyl)methyl]-1,3-thiazolidine-2,4-dione
Synonyms:
rosiglitazone rosiglitazone maleate (substance) glaxosmithkline brand of rosiglitazone maleate smithkline beecham brand of rosiglitazone maleate rosiglitazonum rosiglitazone preparation rosiglitazone (substance) rosiglitazone (product) rosiglizole glaxo wellcome brand of rosiglitazone maleate
Molecular Weight (g/mol):
357.43
Molecular Formula:
C18H19N3O3S
InChi Key:
YASAKCUCGLMORW-UHFFFAOYNA-N
SMILES:
CN(CCOC1=CC=C(CC2SC(=O)NC2=O)C=C1)C1=CC=CC=N1
1
–
5
of
5
results
1
Tocris Bioscience™ Rosiglitazone
Potent and selective PPARγ agonist; antidiabetic agent
2
Rosiglitazone, MedChemExpress
MedChemExpress Rosiglitazone (BRL 49653) is an orally active selective PPARγ agonist (EC50: 60 nM, Kd: 40 nM). Rosiglitazone is an TRPC5 activator (EC50: 30 μM) and TRPM3 inhibitor. Rosiglitazone can be used in the research of obesity and diabetes, senescence, ovarian cancer.
3
4
5