CAS RN 130-86-9
CAS RN 130-86-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Protopine, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
MedChemExpress Protopine
MedChemExpress Protopine, an isoquinoline alkaloid contained in plants in northeast Asia.
3
MedChemExpress Protopine
MedChemExpress Protopine, an isoquinoline alkaloid contained in plants in northeast Asia.