missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protopine, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Protopine |
| CAS | 130-86-9 |
| Analyte or Component Names | Protopine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H19 N O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Bis[1,3]benzodioxolo[4,5-c:5',6'-g]azecin-13(5H)-one, 4,6,7,14-tetrahydro-5-methyl-, 7,13a-Secoberbin-13a-one, 7-methyl-2,3:9,10-bis(methylenedioxy)- (8CI), Protopine (6CI), Biflorine, Corydinine, Fumarin, Fumarine, Macleyine, Protopin |
| InChI Formula | InChI=1S/C20H19NO5/c1-21-5-4-13-7-18-19(25-10-24-18)8-14(13)16(22)6-12-2-3-17-20(15(12)9-21)26-11-23-17/h2-3,7-8H,4-6,9-11H2,1H3 |
| Solution Type | Neat |
| SMILES | CN1CCc2cc3OCOc3cc2C(=O)Cc4ccc5OCOc5c4C1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction