accessibility menu, dialog, popup

UserName

CAS RN 135-19-3

IUPAC Name: naphthalen-2-ol
Synonyms: β naphthol caswell no. 590 2-naphthol, 7-(14)c-labeled 8-(14)c-labeled 2-naphthol (1+) 2-naphthol 7-(14)C-labeled 2-naphthol β-naftol naphthol B titanium(4+) salt 2-naphthol sodium salt 2-naphthol
Molecular Weight (g/mol): 144.17
Molecular Formula: C10H8O
InChi Key: JWAZRIHNYRIHIV-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (13)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (3)
  • (8)
Percent Purity 
  • (3)
  • (3)
  • (5)
Physical Form 
  • (3)
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (8)
  • (3)
  • (3)
  • (13)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (8)
  • (3)
  • (3)
  • (5)
  • (3)
  • (5)
  • (1)