accessibility menu, dialog, popup

UserName

NH3OHNNO

CAS RN 135-20-6

IUPAC Name: N-nitroso-N-phenylhydroxylamine amine
Synonyms: cupferron
Molecular Weight (g/mol): 155.16
Molecular Formula: C6H9N3O2
InChi Key: NJVHCUNZAMFQNA-UHFFFAOYSA-N
SMILES: N.ON(N=O)C1=CC=CC=C1
Quantity 
  • (2)
  • (2)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results