missing translation for 'onlineSavingsMsg'
Learn More

Cupferron

Product Code. 10128040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10128040 500 g 500g
15249636 25 g 25g
10474121 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10128040 Supplier Thermo Scientific Acros Supplier No. 423605000
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 135-20-6 | C6H9N3O2 | 155.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-20-6
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00078422
InChI Key NJVHCUNZAMFQNA-UHFFFAOYSA-N
Synonym cupferron, copperone, azanyliumylidene n-nitroso-n-oxidoaniline
PubChem CID 2724103
IUPAC Name azanium;N-oxido-N-phenylnitrous amide
SMILES N.ON(N=O)C1=CC=CC=C1

Specifications

Melting Point 150.0°C to 155.0°C
Ignition Residue 0.05% max
Packaging Plastic bottle
Linear Formula C6H4N(ONH4)NO
Quantity 500 g
Beilstein 16,I,395
Merck Index 15,2612
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether
Formula Weight 155.16
Grade ACS Reagent
Chemical Name or Material Cupferron
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cupferron
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.