accessibility menu, dialog, popup

UserName

NH2ONHCl

CAS RN 140-38-5

IUPAC Name: (4-chlorophenyl)urea
Synonyms: P-chlorophenylurea
Molecular Weight (g/mol): 170.6
Molecular Formula: C7H7ClN2O
InChi Key: RECCURWJDVZHIH-UHFFFAOYSA-N
SMILES: NC(=O)NC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results