CAS RN 1405-54-5
CAS RN 1405-54-5
IUPAC Name:
(2R,3R)-2,3-dihydroxybutanedioic acid; 2-[(4R,5S,7R,9R,11E,13E,16R)-6-{[(2R,3R,4R,5S,6R)-5-{[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy}-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-16-ethyl-4-hydroxy-15-({[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl]oxy}methyl)-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde
Synonyms:
2-[(4R,5S,7R,9R,11E,13E,16R)-6-{[(2R,3R,4R,5S,6R)-5-{[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy}-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-16-ethyl-4-hydroxy-15-({[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl]oxy}methyl)-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde; L(+)-tartaric acid
Molecular Weight (g/mol):
1066.2
Molecular Formula:
C50H83NO23
InChi Key:
ICVKYYINQHWDLM-YLNALPTESA-N
SMILES:
O[C@H]([C@@H](O)C(O)=O)C(O)=O.CC[C@H]1OC(=O)C[C@@H](O)[C@H](C)C(O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](O)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@@H](CC=O)C[C@@H](C)C(=O)\C=C\C(\C)=C\C1CO[C@@H]1O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]1OC
1
–
1
of
1
results
1
Tylosin tartrate, 94%
CAS: 1405-54-5 Molecular Formula: C50H83NO23 Molecular Weight (g/mol): 1066.198 MDL Number: MFCD00070094 InChI Key: ICVKYYINQHWDLM-HYHSIFHGSA-N Synonym: Pharmasin; [R-(R'R')]-2,3-Dihydroxybutanedionate Tylosin salt PubChem CID: 131673832 IUPAC Name: (2R,3R)-2,3-dihydroxybutanedioic acid;2-[(4R,5S,6S,7R,9R,15R,16R)-6-[(2R,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-16-ethyl-4-hydroxy-15-[[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimetho SMILES: CCC1C(C=C(C=CC(=O)C(CC(C(C(C(CC(=O)O1)O)C)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)O)(C)O)N(C)C)O)CC=O)C)C)COC4C(C(C(C(O4)C)O)OC)OC.C(C(C(=O)O)O)(C(=O)O)O