missing translation for 'onlineSavingsMsg'
Learn More

Tylosin tartrate, 94%

Product Code. 15414969
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414969 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15414969 Supplier Thermo Scientific Alfa Aesar Supplier No. J62633.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Macrolide antibiotic | CAS: 1405-54-5 | C50H83NO23 | 1066.198 g/mol

It is used as an antibacterial agent. This compound has been shown to be effective in vitro against Staphylococcus intermedius and sensitive mycoplasma organisms. Although a lot of antibiotics tend to suppress the immune system, macrolide antibiotics have been reported to enhance the immune system when used in chicken models. Studies suggest that chronic exposure to this compound may have a negative effect on the respiratory system. For turkeys - for treatment of infectious synovitis and sinusitis. It is effective also for treatment of ileitis, dysentery and necrotic enteritis in swine. It is successfully applied as a preventive measure in stress conditions as vaccinations, transportation and others.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an antibacterial agent. This compound has been shown to be effective in vitro against Staphylococcus intermedius and sensitive mycoplasma organisms. Although a lot of antibiotics tend to suppress the immune system, macrolide antibiotics have been reported to enhance the immune system when used in chicken models. Studies suggest that chronic exposure to this compound may have a negative effect on the respiratory system. For turkeys - for treatment of infectious synovitis and sinusitis. It is effective also for treatment of ileitis, dysentery and necrotic enteritis in swine. It is successfully applied as a preventive measure in stress conditions as vaccinations, transportation and others.

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-54-5
Molecular Formula C50H83NO23
Molecular Weight (g/mol) 1066.198
MDL Number MFCD00070094
InChI Key ICVKYYINQHWDLM-HYHSIFHGSA-N
Synonym Pharmasin; [R-(R'R')]-2,3-Dihydroxybutanedionate Tylosin salt
PubChem CID 131673832
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;2-[(4R,5S,6S,7R,9R,15R,16R)-6-[(2R,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-16-ethyl-4-hydroxy-15-[[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimetho
SMILES CCC1C(C=C(C=CC(=O)C(CC(C(C(C(CC(=O)O1)O)C)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)O)(C)O)N(C)C)O)CC=O)C)C)COC4C(C(C(C(O4)C)O)OC)OC.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Type Tylosin Tartrate
Physical Form Powder
Color Cream to White
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 1066.2
Percent Purity 95%
Chemical Name or Material Tylosin tartrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.