accessibility menu, dialog, popup

UserName

CAS RN 142273-20-9

BrNHONH

CAS RN 142273-20-9

IUPAC Name: 14-bromo-8,18-diazatetracyclo[9.7.0.0²,⁷.0¹²,¹⁷]octadeca-1(11),2,4,6,12,14,16-heptaen-9-one
Synonyms: 14-bromo-8,18-diazatetracyclo[9.7.0.0²,⁷.0¹²,¹⁷]octadeca-1(11),2,4,6,12,14,16-heptaen-9-one
Molecular Weight (g/mol): 327.18
Molecular Formula: C16H11BrN2O
InChi Key: QQUXFYAWXPMDOE-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC3=C(CC(=O)NC4=CC=CC=C34)C2=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results