accessibility menu, dialog, popup

UserName

CH3FNOO

CAS RN 1427-07-2

IUPAC Name: 2-fluoro-1-methyl-4-nitrobenzene
Synonyms: 2-fluoro-1-methyl-4-nitrobenzene
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: WIQISTBTOQNVCE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1F)[N+]([O-])=O
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)