missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitrotoluene, 98%

Product Code. 11491778
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491778 5 g 5g
11401788 25 g 25g
11411788 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491778 Supplier Thermo Scientific Alfa Aesar Supplier No. A14464.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1427-07-2 | C7H6FNO2 | 155.128 g/mol

It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.

Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1427-07-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00007199
InChI Key WIQISTBTOQNVCE-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrotoluene, toluene, 2-fluoro-4-nitro, benzene, 2-fluoro-1-methyl-4-nitro, 3-fluoro-4-methylnitrobenzene, 1-methyl-2-fluoro-4-nitrobenzene, 2-fluoro-1-methyl-4-nitro-benzene, pubchem1592, 2-fluoro4-nitrotoluene, acmc-209cov, 2-fluor-4-nitro-toluol
PubChem CID 74025
IUPAC Name 2-fluoro-1-methyl-4-nitrobenzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])F

Specifications

Melting Point 32°C to 37°C
Boiling Point 56°C to 58°C (1 mmHg)
Flash Point 73°C (163°F)
Quantity 5 g
UN Number UN2811
Beilstein 2250156
Solubility Information Insoluble in water. Solubility in methanol gives very faint turbidity.
Formula Weight 155.13
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-nitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.