accessibility menu, dialog, popup

UserName

NH2ClNH2NN

CAS RN 156-83-2

IUPAC Name: 6-chloropyrimidine-2,4-diamine
Synonyms: 6-chloropyrimidine-2,4-diamine
Molecular Weight (g/mol): 144.56
Molecular Formula: C4H5ClN4
InChi Key: QJIUMVUZDYPQRT-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=NC(N)=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results