CAS RN 1616690-16-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Desidustat, TRC
CAS: 1616690-16-4 Molecular Formula: C16 H16 N2 O6 Molecular Weight (g/mol): 332.31 Synonym: N-[[1-(Cyclopropylmethoxy)-1,2-dihydro-4-hydroxy-2-oxo-3-quinolinyl]carbonyl]glycine,ZYAN 1 IUPAC Name: 2-[[1-(cyclopropylmethoxy)-4-hydroxy-2-oxo-quinoline-3-carbonyl]amino]acetic acid SMILES: OC(=O)CNC(=O)C1=C(O)c2ccccc2N(OCC3CC3)C1=O
2
Desidustat, MedChemExpress
MedChemExpress Desidustat is an inhibitor of HIF hydroxylase extracted from patent WO 2014102818 A1, compound example 2.