accessibility menu, dialog, popup

UserName

CAS RN 168555-66-6

H3COOH3COCH3OCH3OPOOHOHNaNa(E/Z)

CAS RN 168555-66-6

IUPAC Name: {2-methoxy-5-[2-(3,4,5-trimethoxyphenyl)ethenyl]phenoxy}phosphonic acid disodium
Synonyms: 2-methoxy-5-[2-(3,4,5-trimethoxyphenyl)ethenyl]phenoxyphosphonic acid disodium
Molecular Weight (g/mol): 442.31
Molecular Formula: C18H21Na2O8P
InChi Key: ACFUENHANVNJPF-UHFFFAOYSA-N
SMILES: [Na].[Na].COC1=CC(C=CC2=CC(OP(O)(O)=O)=C(OC)C=C2)=CC(OC)=C1OC
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results