accessibility menu, dialog, popup

UserName

CH3OOCH3NHN

CAS RN 181122-45-2

IUPAC Name: 5-(2,5-dimethoxyphenyl)-1H-pyrazole
Synonyms: 3-(2,5-dimethoxyphenyl)-2H-pyrazole
Molecular Weight (g/mol): 204.23
Molecular Formula: C11H12N2O2
InChi Key: JEFGLUGSZPRTEJ-UHFFFAOYSA-N
SMILES: COC1=CC(C2=CC=NN2)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results