CAS RN 19398-06-2
IUPAC Name:
(2-ethylphenyl)hydrazine
Synonyms:
(2-ethylphenyl)hydrazine
Molecular Weight (g/mol):
136.2
Molecular Formula:
C8H12N2
InChi Key:
JHPOWXCLWLEKBY-UHFFFAOYSA-N
SMILES:
CCC1=CC=CC=C1NN
1
–
2
of
2
results
1
2-Ethylphenylhydrazine hydrochloride, 97%
CAS: 19398-06-2 Molecular Formula: C8H12N2 Molecular Weight (g/mol): 136.20 MDL Number: MFCD00071599 InChI Key: JHPOWXCLWLEKBY-UHFFFAOYSA-N Synonym: 2-ethylphenylhydrazine hydrochloride,2-ethylphenyl hydrazine hydrochloride,2-ethylphenyl hydrazine hcl,2-ethylphenylhydrazine, hcl,1-2-ethylphenyl hydrazine hydrochloride,2-ethyl phenyl hydrazine hydrochloride,2-ethylphenylhydrazinehydrochloride,hydrazine, 2-ethylphenyl-, monohydrochloride,2-ethylphenylhydrazine, chloride PubChem CID: 2733209 SMILES: CCC1=CC=CC=C1NN
2
2-Ethylphenylhydrazine hydrochloride, TRC
CAS: 19398-06-2 Molecular Formula: C8H13ClN2 Molecular Weight (g/mol): 172.66 SMILES: Cl.CCc1ccccc1NN