missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenylhydrazine hydrochloride, TRC

Product Code. 30381148
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381148 100 mg
30376557 250 mg
30381063 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381148 Supplier Toronto Research Chemicals Supplier No. TRCE926055100MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19398-06-2
Molecular Formula C8H13ClN2
Molecular Weight (g/mol) 172.66
SMILES Cl.CCc1ccccc1NN

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H12N2.ClH/c1-2-7-5-3-4-6-8(7)10-9;/h3-6,10H,2,9H2,1H3;1H
Formula Weight 172.0767
Chemical Name or Material 2-Ethylphenylhydrazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.