missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenylhydrazine hydrochloride, 97%

Product Code. 11482878
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482878 250 g 250g
11462878 10 g 10g
11472878 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482878 Supplier Thermo Scientific Alfa Aesar Supplier No. A16853.30
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 19398-06-2 | C8H12N2 | 136.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19398-06-2
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00071599
InChI Key JHPOWXCLWLEKBY-UHFFFAOYSA-N
Synonym 2-ethylphenylhydrazine hydrochloride, 2-ethylphenyl hydrazine hydrochloride, 2-ethylphenyl hydrazine hcl, 2-ethylphenylhydrazine, hcl, 1-2-ethylphenyl hydrazine hydrochloride, 2-ethyl phenyl hydrazine hydrochloride, 2-ethylphenylhydrazinehydrochloride, hydrazine, 2-ethylphenyl-, monohydrochloride, 2-ethylphenylhydrazine, chloride
PubChem CID 2733209
SMILES CCC1=CC=CC=C1NN

Specifications

Melting Point ∼178°C (decomposition)
Density 1.21
Quantity 250 g
UN Number UN3077
Beilstein 3697547
IUPAC Name (2-ethylphenyl)hydrazine
Formula Weight 172.66
Percent Purity 98%
Chemical Name or Material 2-Ethylphenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.