CAS RN 20427-84-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-Nonyl Phenol Diethoxylate, TRC
CAS: 20427-84-3 Molecular Formula: C19 H32 O3 Molecular Weight (g/mol): 308.46 Synonym: 2-[2-(4-Nonylphenoxy)ethoxy]ethanol,2-[2-(p-Nonylphenoxy)ethoxy]ethanol,4-Nonylphenol diethoxylate,4-n-Nonylphenol diethoxylate,Diethylene glycol mono-p-nonylphenyl ether,Diethylene glycol p-nonylphenyl ether,EON 2 IUPAC Name: 2-[2-(4-nonylphenoxy)ethoxy]ethanol SMILES: CCCCCCCCCc1ccc(OCCOCCO)cc1
2
4-n-Nonylphenol-di-ethoxylate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality