accessibility menu, dialog, popup

UserName

N

CAS RN 2116-65-6

IUPAC Name: 4-benzylpyridine
Synonyms: 4-benzylpyridine
Molecular Weight (g/mol): 169.23
Molecular Formula: C12H11N
InChi Key: DBOLXXRVIFGDTI-UHFFFAOYSA-N
SMILES: C(C1=CC=CC=C1)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)