missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpyridine, TRC

Product Code. 17973665
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973665 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17973665 Supplier Toronto Research Chemicals Supplier No. TRCB2890055G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2116-65-6
Analyte or Component Names 4-Benzylpyridine
Shipping Condition Room Temperature
Molecular Formula C12 H11 N
Synonym 4-Benzylpyridine, Pheniramine Maleate Imp. B (EP)
InChI Formula InChI=1S/C12H11N/c1-2-4-11(5-3-1)10-12-6-8-13-9-7-12/h1-9H,10H2
Solution Type Neat
SMILES C(c1ccccc1)c2ccncc2
IUPAC Name 4-benzylpyridine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 169.22
Quantity 5 g
Formula Weight 169.0891
Chemical Name or Material 4-Benzylpyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.