accessibility menu, dialog, popup

UserName

CAS RN 216393-57-6

BOOH3CCH3FF

CAS RN 216393-57-6

IUPAC Name: 2-(3,5-difluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Synonyms: 2-(3,5-difluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Molecular Weight (g/mol): 226.03
Molecular Formula: C11H13BF2O2
InChi Key: OLHRAEPCGZMMTI-UHFFFAOYSA-N
SMILES: CC1(C)COB(OC1)C1=CC(F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results