CAS RN 2243-06-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Androst-4-ene-3,6,17-trione, TRC
CAS: 2243-06-3 Molecular Formula: C19 H24 O3 Molecular Weight (g/mol): 300.39 Synonym: Androst-4-ene-3,6,17-trione,4-Androstene-3,6,17-trione IUPAC Name: (8R,9S,10R,13S,14S)-10,13-dimethyl-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,6,17-trione SMILES: C[C@]12CC[C@H]3[C@@H](CC(=O)C4=CC(=O)CC[C@]34C)[C@@H]1CCC2=O