accessibility menu, dialog, popup

UserName

CH3OOH3CHO

CAS RN 23287-26-5

IUPAC Name: methyl 2-hydroxy-3-methylbenzoate
Synonyms: methyl 2-hydroxy-3-methylbenzoate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: SUHLUMKZPUMAFP-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(C)=C1O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)