missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxy-3-methylbenzoate, 97%

Product Code. 11449267
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449267 10 g 10g
11439267 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449267 Supplier Thermo Scientific Alfa Aesar Supplier No. B21341.09
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 23287-26-5 | C9H10O3 | 166.18 g/mol

Methyl 2-hydroxy-3-methylbenzoate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic intermediate. Some other applications include it is used to produce other chemicals. For example, it is used to produce 2-(2-Hydroxy-3-methyl-phenyl)-propan-2-ol. The reaction occurs with reagent Tetrahydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-3-methylbenzoate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic intermediate. Some other applications include it is used to produce other chemicals. For example, it is used to produce 2-(2-Hydroxy-3-methyl-phenyl)-propan-2-ol. The reaction occurs with reagent Tetrahydrofuran.

Solubility
Soluble in ethanol, fixed Oils, Propylene Glycol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23287-26-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00020051
InChI Key SUHLUMKZPUMAFP-UHFFFAOYSA-N
Synonym methyl 3-methylsalicylate, methyl o-cresotinate, benzoic acid, 2-hydroxy-3-methyl-, methyl ester, levegal pt, 2-hydroxy-3-methylbenzoic acid methyl ester, 2,3-cresotic acid, methyl ester, methyl-3-methylsalicylate, methyl 2-hydroxy-3-methyl-benzoate, methyl o-cresotate, 3-methylsalicylic acid methyl ester
PubChem CID 31835
IUPAC Name methyl 2-hydroxy-3-methylbenzoate
SMILES COC(=O)C1=CC=CC(C)=C1O

Specifications

Melting Point 28°C to 30°C
Boiling Point 234°C to 236°C
Quantity 10 g
Solubility Information Soluble in ethanol,fixed Oils,Propylene Glycol. Insoluble in water.
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material Methyl 2-hydroxy-3-methylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.