accessibility menu, dialog, popup

UserName

CH3OHH3CH3CCH3

CAS RN 2409-55-4

IUPAC Name: 2-tert-butyl-4-methylphenol
Synonyms: 2-tert-butyl-P-cresol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: IKEHOXWJQXIQAG-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)