missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyl-4-methylphenol, 99%, Thermo Scientific Chemicals

Product Code. 10714791 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10714791 5 g Glass bottle 5g
10214470 100 g Glass bottle 100g
10569990 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10714791 Supplier Thermo Scientific Acros Supplier No. 107960050
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2409-55-4 | C11H16O | 164.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2409-55-4
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00002381
InChI Key IKEHOXWJQXIQAG-UHFFFAOYSA-N
Synonym 2-tert-butyl-4-methylphenol, 2-tert-butyl-p-cresol, 2-t-butyl-p-cresol, p-cresol, 2-tert-butyl, o-tert-butyl-p-cresol, 2-t-butyl-4-methylphenol, 4-methyl-2-tert-butylphenol, 2-terc.butyl-p-kresol, phenol, 2-1,1-dimethylethyl-4-methyl, 4-methyl-6-t-butylphenol
PubChem CID 17004
IUPAC Name 2-tert-butyl-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)C(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 50.0°C to 53.0°C
Color Beige-Yellow to White
Boiling Point 237.0°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H3(CH3)OH
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in aceton, benzene, chloroform
Formula Weight 164.25
Percent Purity 99%
Physical Form Crystalline Mass
Chemical Name or Material 2-tert-Butyl-4-methylphenol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-tert-Butyl-4-methylphenol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.