accessibility menu, dialog, popup

UserName

NaNaNaNaCH3ONHNHNNNSOOOSOOOOSOOOSOOO(E)(E/Z)

CAS RN 2519-30-4

IUPAC Name: tetrasodium (6E)-4-acetamido-5-oxo-6-(2-{7-sulfonato-4-[2-(4-sulfonatophenyl)diazen-1-yl]naphthalen-1-yl}hydrazin-1-ylidene)-5,6-dihydronaphthalene-1,7-disulfonate
Synonyms: brilliant black 1
Molecular Weight (g/mol): 867.66
Molecular Formula: C28H17N5Na4O14S4
InChi Key: AGKKBBSOKGLVTM-YENKNCRGSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].CC(=O)NC1=CC=C(C2=C1C(=O)\C(=N/NC1=C3C=C(C=CC3=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O)S([O-])(=O)=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results