missing translation for 'onlineSavingsMsg'
Learn More

Black PN (~80%), TRC

Product Code. 30134438
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134438 1 g 1g
30132546 10 g 10g
30007428 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134438 Supplier Toronto Research Chemicals Supplier No. TRCB5920001G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2519-30-4
Molecular Formula C28 H17 N5 O14 S4 . 4 Na
Molecular Weight (g/mol) 867.68
Synonym 1,7-Naphthalenedisulfonic acid, 4-(acetylamino)-5-hydroxy-6-[[7-sulfo-4-[(4-sulfophenyl)azo]-1-naphthalenyl]azo]-, tetrasodium salt (9CI), Brilliant Black BN (6CI), C.I. Food Black 1 (7CI), C.I. Food Black 1, tetrasodium salt (8CI), 1743 Black, Black PN, Blue Black BN, Brilliant Acid Black BN Extra Pure A, Brilliant Acid Black BNA Export, Brilliant Black 1, Brilliant Black 80, Brilliant Black A, Brilliant Black N, Brilliant Black NAF, Brilliant Black NFQ, Brilliant Black PN, C.I. 28440, Certicol Black PNW, Cilefa Black B, E 151, Edicol Supra Black BN, Food Black 1, Hexacol Black PN, L Black 8000, Melan Black, Tetrasodium 2-[4-(p-sulfophenylazo)-7-sulfo-1-naphthylazo]-8-acetamido-1-naphthol-3,5-disulfonate, Xylene Black F
IUPAC Name tetrasodium;4-acetamido-5-hydroxy-6-[(E)-[7-sulfonato-4-[(Z)-(4-sulfonatophenyl)diazenyl]naphthalen-1-yl]diazenyl]naphthalene-1,7-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].CC(=O)Nc1ccc(c2cc(c(N=Nc3ccc(N=Nc4ccc(cc4)S(=O)(=O)[O-])c5ccc(cc35)S(=O)(=O)[O-])c(O)c12)S(=O)(=O)[O-])S(=O)(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C28H21N5O14S4.4Na/c1-14(34)29-23-10-11-24(50(42,43)44)20-13-25(51(45,46)47)27(28(35)26(20)23)33-32-22-9-8-21(18-7-6-17(12-19(18)22)49(39,40)41)31-30-15-2-4-16(5-3-15)48(36,37)38;;;;/h2-13,35H,1H3,(H,29,34)(H,36,37,38)(H,39,40,41)(H,42,43,44)(H,45,46,47);;;;/q;4*+1/p-4
Formula Weight 866.9246
Chemical Name or Material Brilliant Black BN
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.