CAS RN 263890-70-6
CAS RN 263890-70-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
GR148672X, MedChemExpress
MedChemExpress GR148672X is a triacyglycerol hydrolase (TGH) inhibitor with an IC50 of 4 nM extracted from patent WO 2001016358 A2.