accessibility menu, dialog, popup

UserName

FFOClFFF

CAS RN 26675-46-7

IUPAC Name: 2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane
Synonyms: isoflurane isoflurano isoflurane (substance) aerrane IsoFlo C3H2ClF5O compound 469 forene forane isofluranum
Molecular Weight (g/mol): 184.49
Molecular Formula: C3H2ClF5O
InChi Key: PIWKPBJCKXDKJR-UHFFFAOYNA-N
SMILES: FC(F)OC(Cl)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results