missing translation for 'onlineSavingsMsg'
Learn More

Isoflurane, Mikromol™

Product Code. 30049371
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049371 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 30049371 Supplier Mikromol Supplier No. MM0500.000250
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Isoflurane
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isoflurane
CAS 26675-46-7
Analyte or Component Names Isoflurane
Shipping Condition Ice pack (-20°C)
Molecular Formula C3 H2 Cl F5 O
Certifications/Compliance ISO 9001
Synonym (2RS)-2-Chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane, Ether, 1-chloro-2,2,2-trifluoroethyl difluoromethyl (8CI), 2-Chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane, (±)-Isoflurane, 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether, Aerrane, Attane, Compound 469, Difluoromethyl 1-chloro-2,2,2-trifluoroethyl ether, Escain, Forane, Forene, HCFE 235da2, IsoFlo, IsoFlo (anesthetic), Isoba, Isoflurane, Isofor, Isoforine, Isorrane, R-E 235da1
InChI Formula InChI=1S/C3H2ClF5O/c4-1(3(7,8)9)10-2(5)6/h1-2H
SMILES FC(F)OC(Cl)C(F)(F)F
IUPAC Name 2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane
Molecular Weight (g/mol) 184.49
Quantity 250 mg
Formula Weight 183.9714 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.