accessibility menu, dialog, popup

UserName

CH3NH2HOO

CAS RN 2941-78-8

IUPAC Name: 2-amino-5-methylbenzoic acid
Synonyms: 2-amino-5-methylbenzoic acid
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: NBUUUJWWOARGNW-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results