missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylbenzoic acid, 97%

Product Code. 11341307
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11341307 25 g 25g
11321307 1 g 1g
11331307 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11341307 Supplier Thermo Scientific Alfa Aesar Supplier No. L01513.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2941-78-8 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2941-78-8
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007909
InChI Key NBUUUJWWOARGNW-UHFFFAOYSA-N
Synonym 5-methylanthranilic acid, 6-amino-m-toluic acid, 2-amino-5-methyl-benzoic acid, benzoic acid, 2-amino-5-methyl, m-toluic acid, 6-amino, 2-amino-5-methylbenzoicacid, unii-418xaj22v2, 2-amino-5-methyl benzoic acid, amino 2--5-methylbenzoic acid, pubchem4954
PubChem CID 76255
IUPAC Name 2-amino-5-methylbenzoic acid
SMILES CC1=CC=C(N)C(=C1)C(O)=O

Specifications

Melting Point ∽175°C (decomposition)
Quantity 25 g
Beilstein 1101527
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 2-Amino-5-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.