missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylbenzoic acid, 97%

Product Code. 10470432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10470432 5 g Glass bottle 5g
10460432 1 g Glass bottle 1g
10299863 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10470432 Supplier Thermo Scientific Acros Supplier No. 160200050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2941-78-8 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2941-78-8
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007909
InChI Key NBUUUJWWOARGNW-UHFFFAOYSA-N
Synonym 5-methylanthranilic acid, 6-amino-m-toluic acid, 2-amino-5-methyl-benzoic acid, benzoic acid, 2-amino-5-methyl, m-toluic acid, 6-amino, 2-amino-5-methylbenzoicacid, unii-418xaj22v2, 2-amino-5-methyl benzoic acid, amino 2--5-methylbenzoic acid, pubchem4954
PubChem CID 76255
IUPAC Name 2-amino-5-methylbenzoic acid
SMILES CC1=CC=C(N)C(=C1)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 174.0°C to 177.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(CH3)CO2H
Quantity 5 g
Beilstein 14, 481
Formula Weight 151.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-5-methylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-methylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.