accessibility menu, dialog, popup

UserName

NH2FFF

CAS RN 3048-01-9

IUPAC Name: 1-[2-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[2-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 175.15
Molecular Formula: C8H8F3N
InChi Key: ZSKQIFWUTUZAGF-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results