missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzylamine, 98%, Thermo Scientific™

Product Code. 11480288
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480288 10 g 10g
11470288 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480288 Supplier Thermo Scientific Alfa Aesar Supplier No. B23822.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 3048-01-9
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.15
MDL Number MFCD00010297
InChI Key ZSKQIFWUTUZAGF-UHFFFAOYSA-N

Specifica

Density 1.249
Boiling Point 108°C to 110°C (64 mmHg)
Flash Point 69°C (156°F)
Refractive Index 1.471
Quantity 10 g
UN Number UN2735
Beilstein 1103424
Sensitivity Air Sensitive
Synonym direct yellow 4, brilliant yellow, paper yellow cb, unii-zun7l1adie, zun7l1adie, brilliant yellow c, paper yellow 3gx, fenamin yellow 3g, paper yellow 3gxa, c.i. direct yellow 4, disodium salt
SMILES NCC1=CC=CC=C1C(F)(F)F
IUPAC Name 1-[2-(trifluoromethyl)phenyl]methanamine
PubChem CID 5702677
Formula Weight 175.15
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)benzylamine
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.