accessibility menu, dialog, popup

UserName

CAS RN 30980-37-1

OO

CAS RN 30980-37-1

IUPAC Name: [(6-phenoxyhexa-2,4-diyn-1-yl)oxy]benzene
Synonyms: [(6-phenoxyhexa-2,4-diyn-1-yl)oxy]benzene
Molecular Weight (g/mol): 262.31
Molecular Formula: C18H14O2
InChi Key: ZBTRFBNXAKJTBI-UHFFFAOYSA-N
SMILES: C(OC1=CC=CC=C1)C#CC#CCOC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results