accessibility menu, dialog, popup

UserName

H3COOHOHO

CAS RN 3147-64-6

IUPAC Name: 2-hydroxy-6-methoxybenzoic acid
Synonyms: 2-hydroxy-6-methoxybenzoic acid
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: AAUQLHHARJUJEH-UHFFFAOYSA-N
SMILES: COC1=CC=CC(O)=C1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)