missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxysalicylic acid, 98%, Thermo Scientific™

Product Code. 10821891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10821891 10 g 10g
10250220 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10821891 Supplier Thermo Scientific Acros Supplier No. 437670100
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3147-64-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
InChI Key AAUQLHHARJUJEH-UHFFFAOYSA-N
Synonym 6-methoxysalicylic acid, 6-hydroxy-o-anisic acid, benzoic acid, 2-hydroxy-6-methoxy, 6-methoxysalicylicacid, 6-methoxysalicylsaure, pubchem18380, acmc-1cldg, 6-methoxy-salicylic acid, 2-carboxy-3-methoxyphenol, 2-carboxy-3-hydroxyanisole
PubChem CID 591524
IUPAC Name 2-hydroxy-6-methoxybenzoic acid
SMILES COC1=CC=CC(=C1C(=O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 135.9°C to 136.4°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 10 g
Formula Weight 168.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-Methoxysalicylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methoxysalicylic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.