accessibility menu, dialog, popup

UserName

CAS RN 320-31-0

BrFFFOHO

CAS RN 320-31-0

IUPAC Name: 4-bromo-2-(trifluoromethyl)benzoic acid
Synonyms: 4-bromo-2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 269.02
Molecular Formula: C8H4BrF3O2
InChi Key: JXHWAPDBDXPBEQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=C(Br)C=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results