missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethyl)benzoic acid, 98%

Product Code. 15414267
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414267 5 g 5g
15404267 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414267 Supplier Thermo Scientific Alfa Aesar Supplier No. H31656.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 320-31-0 | C8H4BrF3O2 | 269.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 320-31-0
Molecular Formula C8H4BrF3O2
Molecular Weight (g/mol) 269.02
MDL Number MFCD07787468
InChI Key JXHWAPDBDXPBEQ-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethyl benzoic acid, benzoic acid, 4-bromo-2-trifluoromethyl, 2-trifluoromethyl-4-bromobenzoic acid, pubchem4738, 4-bromo-2-trifluoromethyl benzoicacid, acmc-209hq4, ksc495g7n, 5-bromo-2-carboxybenzotrifluoride
PubChem CID 25067392
IUPAC Name 4-bromo-2-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=C(C=C(Br)C=C1)C(F)(F)F

Especificaciones

Quantity 5 g
Formula Weight 269.02
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)benzoic acid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.